CAS RN 2018282-44-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
BMT-145027, MedChemExpress
MedChemExpress BMT-145027 is an mGluR5 positive allosteric modulator without inherent agonist activity, exhibits an EC50 of 47 nM.