CAS RN 201410-67-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
(-)-Talarozole, MedChemExpress
MedChemExpress (-)-Talarozole is a potent inhibitor of retinoic acid metabolism extracted from patent WO 1997049704 A1.