CAS RN 2003234-64-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
(S)-GNE-140, MedChemExpress
MedChemExpress (S)-GNE-140 is the less active enantiomer of GNE-140 which can inhibit Lactate dehydrogenase A (LDHA).