CAS RN 200132-83-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Kasugamycin hydrochloride hydrate, MedChemExpress
MedChemExpress Kasugamycin hydrochloride hydrate (Ksg hydrochloride hydrate) is an antibiotic which binds both the 30S and 70S ribosome but not isolated 50S subunits. Kasugamycin hydrochloride hydrate (Ksg hydrochloride hydrate) mimics mRNA nucleotides to destabilize tRNA binding and inhibit canonical translation initiation.