CAS RN 19970-45-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
SJ572403, MedChemExpress
MedChemExpress SJ572403 (SJ403) is an inhibitor of disordered protein p27(Kip1). p27(Kip1) is a regulator of the CDKs that control eukaryotic cell division.