CAS RN 199293-83-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
MedChemExpress NSP-SA-NHS
MedChemExpress NSP-SA-NHS is an acridinium ester that can be used for chemiluminescent immunoassay. A rapid and sensitive chemiluminescent immunoassay of Bisphenol A with NSP-SA-NHS-labeled has been developed.