CAS RN 197894-84-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
CI-1044, MedChemExpress
MedChemExpress CI-1044 is an orally active PDE4 inhibitor with IC50s of 0.29, 0.08, 0.56, 0.09 μM for PDE4A5, PDE4B2, PDE4C2 and PDE4D3, respectively.