CAS RN 19608-29-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Clascoterone, MedChemExpress
MedChemExpress Clascoterone (Cortexolone 17 alpha-propionate;Cortexolone 17α-propionate;CB-03-01) is a new topical and peripherally selective androgen antagonist.