CAS RN 1953146-81-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Tocris Bioscience™ tri-GalNAc COOH
Asialoglycoprotein receptor ligand with PEG linker and carboxylic acid group for onward chemistry