CAS RN 1950635-16-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Thalidomide-O-amido-C8-NH2 TFA, MedChemExpress
MedChemExpress Thalidomide-O-amido-C8-NH2 TFA (Cereblon Ligand -Linker Conjugates 2 TFA) is a synthesized E3 ligase ligand-linker conjugate that incorporates the Thalidomide based cereblon ligand and a linker used in PROTAC technology.