CAS RN 1943733-16-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
YU238259, MedChemExpress
MedChemExpress YU238259 is an inhibitor of homology-dependent DNA repair (HDR), used for cancer research.