CAS RN 1940176-03-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Abiraterone metabolite 1, MedChemExpress
MedChemExpress Abiraterone metabolite 1 is a 5β-reduced metabolite of abiraterone. Abiraterone, a steroidal drug, inhibits CYP17A1, blocks androgen synthesis and prolongs survival in prostate cancer.