CAS RN 192203-60-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Carboxypeptidase G2 (CPG2) Inhibitor, MedChemExpress
MedChemExpress Carboxypeptidase G2 (CPG2) Inhibitor is a novel Carboxypeptidase G2 (CPG2) Inhibitor, Antitumor agents.