CAS RN 189224-26-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Ozagrel sodium, MedChemExpress
MedChemExpress Ozagrel sodium (OKY-046 sodium) is a thromboxane A2 (TXA2) synthase inhibitor. Ozagrel sodium is an antiplatelet agent, which selectively inhibits human platelet aggregation with an IC50 of 53.12 μM.