CAS RN 1884712-47-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
CPI-637, MedChemExpress
MedChemExpress CPI-637 is a selective and potent CBP/EP300 bromodomain inhibitor with IC50 values of 0.03 μM, 0.051 μM and 11.0 μM for CBP, EP300 and BRD4 BD-1, respectively, and an EC50 of 0.3 μM for CBP.