CAS RN 1883727-34-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Tapotoclax, MedChemExpress
MedChemExpress Tapotoclax (AMG-176) is a potent, selective and orally active MCL-1 inhibitor, with a Ki of 0.13 nM.