CAS RN 1883429-21-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
BI-7273, MedChemExpress
MedChemExpress BI-7273 is a selective, and cell-permeable BRD9 inhibitor, with an IC50 and a Kd of 19 and 0.75 nM; also shows high effect on BRD7, with an IC50 and a Kd of 117 nM and 0.3 nM.