CAS RN 1862238-00-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Sirt2-IN-1, MedChemExpress
MedChemExpress Sirt2-IN-1 (Compound 9) is a sirtuin 2 (Sirt2) inhibitor with an IC50 of 163 nM.