CAS RN 186185-03-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
BMS-193885, MedChemExpress
MedChemExpress BMS-193885 is a potent, selective, competitive, and brain penetrant neuropeptide Y1 receptor antagonist with a Ki of 3.3 nM, and has an IC50 of 5.9 nM for hY1, which displays > 100, > 160, > 160 and > 160-fold selectivity over α1, hY2, hY4 and hY5 receptors, respectively .