CAS RN 1857417-13-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
MI-503, MedChemExpress
MedChemExpress MI-503 is a highly potent and orally bioavailable small molecule inhibitor of the menin-mLL interaction.