CAS RN 185213-52-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Scabertopin, MedChemExpress
MedChemExpress Scabertopin is a sesquiterpene lactone. Scabertopin has been found to be prominent anticancer constituents.