CAS RN 1835705-53-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
MedChemExpress Boc-C1-PEG2-C4-Cl
MedChemExpress Boc-C1-PEG2-C4-Cl (PROTAC Linker 1) is a PEG-based PROTAC linker can be used in the synthesis of PROTACs.