CAS RN 1835705-52-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Boc-C5-O-C5-O-C6-Cl, MedChemExpress
MedChemExpress Boc-C5-O-C5-O-C6-Cl (PROTAC Linker 2) is a PROTAC linker utilized to connect the respective tyrosine kinase inhibitor (TKI) to the E3 recruiting ligand.