CAS RN 18356-28-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Rolziracetam, MedChemExpress
MedChemExpress Rolziracetam is a nootropic drug of the racetam family and improves short-term memory in rats and monkeys.