CAS RN 1811539-37-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Tocris Bioscience™ Janelia Fluor™ 525, Haloalkane
Cell-permeable fluorogenic fluorescent dye with a chloroalkane handle. Application: live-cell imaging as a self-labeling tag substrate