CAS RN 1808260-84-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
FASN-IN-1, MedChemExpress
MedChemExpress FASN-IN-1 is a fatty acid synthase (FASN) inhibitor extracted from patent WO2015134790A1, compound 56.