CAS RN 1808094-07-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
PF-06471553, MedChemExpress
MedChemExpress PF-06471553 is a potent, selective and orally available monoacylglycerol acyltransferase 3 (MGAT3) inhibitor, with an IC50 of 92 nM.