CAS RN 1802525-61-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
LRRK2 inhibitor 1, MedChemExpress
MedChemExpress LRRK2 inhibitor 1 is a potent, selective and oral LRRK2 inhibitor with an pIC50 of 6.8.