CAS RN 180084-26-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Tocris Bioscience™ SB 224289 hydrochloride
Selective 5-HT1B antagonist
2
SB-224289 hydrochloride, MedChemExpress
MedChemExpress SB-224289 hydrochloride is a selective 5-HT1B receptor antagonist, with anxiolytic effect.