CAS RN 179528-45-1
IUPAC Nombre:
3-(4-iodophenyl)-2-sulfanylprop-2-enoic acid
Sinónimos:
3-(4-iodophenyl)-2-sulfanylprop-2-enoic acid
Peso molecular (g/mol):
306.12
Fórmula molecular:
C9H7IO2S
InChi Key:
DJCVSFWGKYHMKH-UHFFFAOYSA-N
SMILES:
OC(=O)C(S)=CC1=CC=C(I)C=C1
1
–
2
de
2
resultados
1
PD150606, MedChemExpress
MedChemExpress PD 150606 is a selective, cell-permeable non-peptide calpain inhibitor with Ki values of 0.21 μM and 0.37 μM for μ- and m-calpains respectively, which is neuroprotective.
2
Tocris Bioscience™ PD 150606
CAS: 179528-45-1 Fórmula molecular: C9H7IO2S