CAS RN 179068-02-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
PHCCC, MedChemExpress
MedChemExpress PHCCC is a Group I mGluR antagonist with an IC50 of 3 μM. PHCCC is a selective positive modulator of mGlu4 receptor. Antiparkinsonian effect.