CAS RN 1781835-02-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
Tocris Bioscience™ WAY 316606 hydrochloride
Secreted Frizzled-related protein-1 (sFRP-1) inhibitor; increases Wnt signaling
2
Tocris Bioscience™ WAY 316606 hydrochloride
Secreted Frizzled-related protein-1 (sFRP-1) inhibitor; increases Wnt signaling
3
Tocris Bioscience™ WAY 316606 hydrochloride
Secreted Frizzled-related protein-1 (sFRP-1) inhibitor; increases Wnt signaling