CAS RN 175131-60-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Milnacipran ((1S-cis) hydrochloride), MedChemExpress
MedChemExpress Milnacipran (1S-cis) hydrochloride is a serotonin-norepinephrine reuptake inhibitor (SNRI), used in the clinical treatment of fibromyalgia.