CAS RN 174635-69-9
IUPAC Nombre:
3-methyl-2-phenyl-N-(1-phenylpropyl)quinoline-4-carboxamide
Sinónimos:
3-methyl-2-phenyl-N-(1-phenylpropyl)quinoline-4-carboxamide
Peso molecular (g/mol):
380.49
Fórmula molecular:
C26H24N2O
InChi Key:
MQNYRKWJSMQECI-UHFFFAOYNA-N
SMILES:
CCC(NC(=O)C1=C2C=CC=CC2=NC(=C1C)C1=CC=CC=C1)C1=CC=CC=C1
1
–
2
de
2
resultados
1
Tocris Bioscience™ SB 222200
CAS: 174635-69-9 Fórmula molecular: C26H24N2O
2
SB-222200, MedChemExpress
MedChemExpress SB-222200 is a potent, selective, orally active and blood-brain barrier (BBB) penetrant NK-3 receptor antagonist. SB-222200 is developed for central nervous system (CNS) disorders.