CAS RN 171436-38-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
MedChemExpress SLIGRL-NH2
MedChemExpress SLIGRL-NH2 (Protease-Activated Receptor-2 Activating Peptide) is an agonist of Protease-Activated Receptor-2 (PAR-2).
2
MedChemExpress SLIGRL-NH2
MedChemExpress SLIGRL-NH2 (Protease-Activated Receptor-2 Activating Peptide) is an agonist of Protease-Activated Receptor-2 (PAR-2).