CAS RN 1708971-55-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Roblitinib, MedChemExpress
MedChemExpress Roblitinib (FGF-401) is an orally active and highly selective FGFR4 inhibitor with an IC50 of 1.9 nM. Roblitinib has antitumor activity.