CAS RN 170632-47-0
IUPAC Nombre:
[5-(1-benzyl-1H-indazol-3-yl)furan-2-yl]methanol
Sinónimos:
lificiguat
Peso molecular (g/mol):
304.35
Fórmula molecular:
C19H16N2O2
InChi Key:
OQQVFCKUDYMWGV-UHFFFAOYSA-N
SMILES:
OCC1=CC=C(O1)C1=NN(CC2=CC=CC=C2)C2=CC=CC=C12
1
–
2
de
2
resultados
1
Tocris Bioscience™ YC 1
CAS: 170632-47-0 Fórmula molecular: C19H16N2O2
2
Lificiguat, MedChemExpress
MedChemExpress Lificiguat binds to the β subunit of soluble guanylyl cyclase(sGC) with Kd of 0.6-1.1 μM in the presence of CO.