CAS RN 170006-72-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
FTI 276, MedChemExpress
MedChemExpress FTI-276 is a protein farnesyl transferase (PFT) inhibitor with IC50s of 0.9 and 0.5 nM for Plasmodium falciparum and human.