CAS RN 16994-56-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Reserpine hydrochloride, MedChemExpress
MedChemExpress Reserpine hydrochloride is an inhibitor of the vesicular monoamine transporter 2 (VMAT2).