CAS RN 16947-80-1
IUPAC Nombre:
2-{[(tert-butoxy)carbonyl]amino}-3-(methylsulfanyl)propanoic acid
Sinónimos:
2-[(tert-butoxycarbonyl)amino]-3-(methylsulfanyl)propanoic acid
Peso molecular (g/mol):
235.3
Fórmula molecular:
C9H17NO4S
InChi Key:
WFDQTEFRLDDJAM-UHFFFAOYNA-N
SMILES:
CSCC(NC(=O)OC(C)(C)C)C(O)=O
1
–
1
de
1
resultados
1
N-Boc-S-metil-L-cisteína, 96 %, Thermo Scientific Chemicals
CAS: 16947-80-1 Fórmula molecular: C9H17NO4S Peso molecular (g/mol): 235.30 Número MDL: MFCD00076923 Clave InChI: WFDQTEFRLDDJAM-UHFFFAOYNA-N Sinónimo: n-boc-s-methyl-l-cysteine,boc-cys me-oh,boc-s-methylcysteine,boc-s-methyl-l-cysteine,2r-2-tert-butoxycarbonyl amino-3-methylsulfanyl propanoic acid,l-cysteine, n-1,1-dimethylethoxy carbonyl-s-methyl,boccys me oh,cysteine, n-1,1-dimethylethoxy carbonyl-s-methyl,n-boc-s-methyl-l-cys-oh,r-boc-s-methyl cysteine PubChem CID: 7018744 Nombre IUPAC: ácido (2R)-2-[(2-metilpropan-2-il)oxicarbonilamino]-3-metilsulfanilpropanoico SMILES: CSCC(NC(=O)OC(C)(C)C)C(O)=O