CAS RN 168635-85-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
MedChemExpress Carcinoembryonic Antigen (CEA)
MedChemExpress Carcinoembryonic Antigen (CEA) is a tumor marker in lung cancer.
2
MedChemExpress Carcinoembryonic Antigen (CEA)
MedChemExpress Carcinoembryonic Antigen (CEA) is a tumor marker in lung cancer.