CAS RN 166734-83-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Ornidazole (Levo-), MedChemExpress
MedChemExpress Ornidazole Levo- is the levo-isomer of Ornidazole. Ornidazole is a 5-nitroimidazole derivative with antiprotozoal and antibacterial properties against anaerobic bacteria. Ornidazole Levo- is the less active isomer.