CAS RN 16662-47-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Gallopamil, MedChemExpress
MedChemExpress Gallopamil (Methoxyverapamil), a methoxy derivative of Verapamil, is a phenylalkylamine calcium antagonist. Gallopamil inhibits acid secretion in a concentration-dependent manner with an IC50 of 10.9 μM. Gallopamil is a potent antiarrhythmic and vasodilator agent.