CAS RN 1661839-45-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
CCT251545, MedChemExpress
MedChemExpress CCT251545 is an orally bioavailable and potent inhibitor of WNT signaling with an IC50 of 5 nM in 7dF3 cells. CCT251545 is a selective chemical probe for exploring the role of CDK8 and CDK19 in human disease.