CAS RN 16611-84-0
IUPAC Nombre:
2-hydroxy-6-pentadecylbenzoic acid
Sinónimos:
anacardic acid
Peso molecular (g/mol):
348.53
Fórmula molecular:
C22H36O3
InChi Key:
ADFWQBGTDJIESE-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCC1=CC=CC(O)=C1C(O)=O
1
–
2
de
2
resultados
1
Anacardic Acid, MedChemExpress
MedChemExpress Anacardic Acid, extracted from cashew nut shell liquid, is a histone acetyltransferase inhibitor, inhibits HAT activity of p300 and PCAF, with IC50s of ∼8.5 μM and ∼5 μM, respectively.
2
Tocris Bioscience™ Anacardic acid
CAS: 16611-84-0 Fórmula molecular: C22H36O3