CAS RN 1644545-52-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Vorasidenib, MedChemExpress
MedChemExpress Vorasidenib (AG-881) is an orally available, brain penetrant second-generation dual mutant isocitrate dehydrogenases 1 and 2 (mIDH1/2) inhibitor. Vorasidenib (AG-881) exhibits nanomolar inhibition of (D)-2-hydroxyglutarate (D-2-HG), and the IC50 ranges of 0.04∼22 nM against IDH1 R132C, IDH1 R132G, IDH1 R132H and IDH1 R132S and 7∼14 nM against IDH2 R140Q and 130 nM against IDH2 R172K.