CAS RN 1642857-69-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
2
TLR7 agonist 2, MedChemExpress
MedChemExpress TLR7 agonist 2 is a potent and selective Toll-like Receptor 7 (TLR7) agonist with a LEC of 0.4 μM.