CAS RN 1639895-85-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
tBID, MedChemExpress
MedChemExpress tBID is a selective inhibitor of homeodomain-interacting protein kinase 2 (HIPK2) with an IC50 of 0.33 μM.