CAS RN 163706-36-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Tocris Bioscience™ AR-C 69931 Tetrasodium Salt
Low molecular weight, organic compound that may help regulate biological process.