CAS RN 1633350-06-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
(±)-Zanubrutinib, MedChemExpress
MedChemExpress (±)-Zanubrutinib ((±)-BGB-3111) is a potent, selective and orally available Bruton's tyrosine kinase (Btk) inhibitor.